Learn More
AVANTOR MASTERFLEX (VWR LLC) DIGITAL BENCHTOP GEAR PUMP SYS
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7401442UK
Encompass
Masterflex Digital Benchtop Gear Pump System, 1.17 mL/rev, 60 psi; 90 to 260 VAC
Catalog No. NC2280508