Learn More
Vernier™ Breadboard Cable (Digital)
SDPSCI_ED
Catalog No. S16013ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With: Interfaces
Catalog No.
S16013ND For Use With: Interfaces
1 options
Catalog No. S16013ND Supplier Vernier™ Supplier No. BBBTD
Add to Cart
Add to Cart
Includes
Breadboard Cable (Digital)
The Breadboard Cable (Digital) provides an easy way for students to build their own sensor circuitry and input the signal into a Vernier interface for data acquisition.
  • One end of the cable is a standard British Telecom Digital (BTD) connector that plugs into a compatible interface.
  • he other end of the cable is a prototyping board connector that exposes the six sensor lines on a prototyping board.
Warranty and Services
5-year limited warranty
Specifications
Product Type Digital
For Use With (Equipment) Interfaces
For Use With (Application) Educational use only
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov