Learn More
Andwin Scientific DIGITAL CALIPER
SDP
Catalog No. NC9719599
Encompass_Preferred
The Digital Calipers are made to measure the outside diameter of stoppers, inside diameter of tubing, and depth of vials. A zero button is present to allow quick set of closed jaws or any reference point to zero. The Floating Zero™ may also be set in
Catalog No. NC9719599 Supplier Andwin Scientific Supplier No. 62379531
The Digital Calipers are made to measure the outside diameter of stoppers, inside diameter of tubing, and depth of vials. A zero button is present to allow quick set of closed jaws or any reference point to zero. The Floating Zero™ may also be set in