Learn More
Fine Science Tools Digital Electronic Caliper
SDP
Catalog No. NC1926306
Encompass
Digital Electronic Caliper, Increments: 0.1mm, Maximum Measurement: 152.4mm, Wide screen LED display with the ability to measure in millimeters, inches, and fractions. Measure inside, outside, step and depth. Battery, spare battery and plastic storage case included.
Catalog No. NC1926306 Supplier Fine Science Tools Supplier No. 3008700
Digital Electronic Caliper, Increments: 0.1mm, Maximum Measurement: 152.4mm, Wide screen LED display with the ability to measure in millimeters, inches, and fractions. Measure inside, outside, step and depth. Battery, spare battery and plastic storage case included.