Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotators

Catalog No. 88882016
Change view
Click to view available options
Plug Type:
EU, UK, CN Plug
US Plug
Certifications/Compliance:
CE
cCSAus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Certifications/Compliance
88-882-016 EU, UK, CN Plug CE
88-882-015 US Plug cCSAus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 88-882-016 Supplier Thermo Scientific™ Supplier No. 88882016
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Requires

Minimum 1 drum order together with Rotator

The Digital Cel-Gro Tissue Culture Rotator has a brushless no-maintenance motor that provides a gentle rotating motion for optimum results.

The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.

  • Digital display of Speed and Time
  • Speed range: 5 – 70 rpm
  • Speed Accuracy: ±1 rpm
  • Adjustable rocking Angle: 95°- 180°
  • Loading Capacity: 5 kg
  • Timer Range: 0 - 9999mins
  • Optional multiple size drums offering different capacity of tube rotation

Order Info

Please order minimum 1 drum together with the rotator

Warranty and Services

2 years

TRUSTED_SUSTAINABILITY

Specifications

Load Bearing Capacity (Metric) 5 kg
Speed Control Accuracy ±1 rpm
Tilt Angle 95 to 180°
Speed Range 5 to 70 rpm
Electrical Requirements 100/240 V
Amperage 0.2 A
Certifications/Compliance CE
Plug Type EU, UK, CN Plug
Temperature Range (English) 5-40°F
Frequency 50/60 Hz
Material Stainless Steel Platform
Dimensions (L x W x H) 383 x 260 x 522 mm (15.1 x 10.2 x 20.6 in.)
Shipping Weight (English) 26 lb.
Shipping Weight (Metric) 11.8 kg
Alarms Audible alarm
Controller Type Digital control
Model Digital Cel-Gro Tissue Culture Rotator
Display Digital
For Use With (Application) Cell Culture
For Use With (Equipment) Accessories: 88882149, 88882150,88882151, 88882152, 88882153, 88882154, 88882155, 88882156, 88882157
Humidity Range 20 to 85%
Timer Range 0 to 9999 min.
Warranty and Service Offering 2 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.