Learn More
MSC General Tools and Instruments Hand-Held Carbon Monoxide Meter, 0 to 999 ppm, Backlit LCD, 1 ea
Catalog No. 5000688371
Encompass
This hand-held digital carbon monoxide (CO) meter provides accurate monitoring for homes, offices, and industrial environments. It utilizes an electrochemical sensor to deliver reliable detection and features a dual-readout display for real-time monitoring and maximum hold values.
  • Measurement range of 0 ppm to 999 ppm with 1 ppm resolution
  • Backlit dual LCD displays real-time CO levels and maximum values
  • Adjustable audible alarm thresholds starting at 25 ppm
  • One-button calibration for 0 ppm, 100 ppm, or 500 ppm
  • Fifteen-second self-test at startup
  • Auto power off after 15 minutes of inactivity
  • Includes three AAA batteries, wrist strap, carrying pouch, and manual
  • Operates on three AAA batteries with up to 250 hours of life
Catalog No. 50-006-88371 Supplier MSC Supplier No. 93811958
This hand-held digital carbon monoxide (CO) meter provides accurate monitoring for homes, offices, and industrial environments. It utilizes an electrochemical sensor to deliver reliable detection and features a dual-readout display for real-time monitoring and maximum hold values.
  • Measurement range of 0 ppm to 999 ppm with 1 ppm resolution
  • Backlit dual LCD displays real-time CO levels and maximum values
  • Adjustable audible alarm thresholds starting at 25 ppm
  • One-button calibration for 0 ppm, 100 ppm, or 500 ppm
  • Fifteen-second self-test at startup
  • Auto power off after 15 minutes of inactivity
  • Includes three AAA batteries, wrist strap, carrying pouch, and manual
  • Operates on three AAA batteries with up to 250 hours of life