Learn More
MSC Mahr 30 EWRi-N Electronic Depth Gage, 0-100 mm, Stainless Steel, IP67, 1 ea
Catalog No. 5000512227
Encompass
These electronic depth gages provide accurate measurements for industrial applications, featuring a 0 to 100 mm (0 to 4 in.) measurement range and a resolution of 0.01 mm (0.0005 in.). Constructed with hardened stainless steel, the instrument includes a lapped guideway and integrated dirt wipers for enhanced durability and resistance to dust, coolants, and lubricants. It is designed for immediate measurements via an integrated reference system and features a high-contrast digital display.
  • Range: 0 to 100 mm (0 to 4 in.)
  • Accuracy: 0.03 mm (0.001 in.)
  • IP 67 protection rating
  • Hardened stainless steel slide and beam
  • Data transmission via USB, Opto RS-232C, or Digimatic
  • Battery-powered with approximately 3-year lifespan
  • Includes battery, case, instruction manual, and anvils
Catalog No. 50-005-12227 Supplier MSC Supplier No. 50486232
These electronic depth gages provide accurate measurements for industrial applications, featuring a 0 to 100 mm (0 to 4 in.) measurement range and a resolution of 0.01 mm (0.0005 in.). Constructed with hardened stainless steel, the instrument includes a lapped guideway and integrated dirt wipers for enhanced durability and resistance to dust, coolants, and lubricants. It is designed for immediate measurements via an integrated reference system and features a high-contrast digital display.
  • Range: 0 to 100 mm (0 to 4 in.)
  • Accuracy: 0.03 mm (0.001 in.)
  • IP 67 protection rating
  • Hardened stainless steel slide and beam
  • Data transmission via USB, Opto RS-232C, or Digimatic
  • Battery-powered with approximately 3-year lifespan
  • Includes battery, case, instruction manual, and anvils