Learn More
MSC Insize Electronic Protractor, 360 Degree, 0.1 Degree Resolution, 1 ea
Catalog No. NC2008458
Encompass
This electronic protractor is designed for precise angle measurements, featuring a 0-360 degree measuring range and a high resolution of 0.1 degrees. It is suitable for various applications requiring accurate angular readings.
  • Electronic display with on/off and zero functions for ease of use
  • Clamping screw to lock the blades for stable measurements
  • Blade with graduations of 1 mm and 1/32 in for versatile linear measurement
  • Constructed from stainless steel for durability and longevity
  • Powered by a CR2032 battery with an automatic power-off function to conserve power
  • Includes a protective case for storage and transport
Catalog No. NC2008458 Supplier MSC Supplier No. 96548847
This electronic protractor is designed for precise angle measurements, featuring a 0-360 degree measuring range and a high resolution of 0.1 degrees. It is suitable for various applications requiring accurate angular readings.
  • Electronic display with on/off and zero functions for ease of use
  • Clamping screw to lock the blades for stable measurements
  • Blade with graduations of 1 mm and 1/32 in for versatile linear measurement
  • Constructed from stainless steel for durability and longevity
  • Powered by a CR2032 battery with an automatic power-off function to conserve power
  • Includes a protective case for storage and transport