Learn More
Medicus Health Digital Differential Counter
SDP
Catalog No. NC3302221
Encompass
Digital Counting Device is an advanced tool designed for accurate and efficient white blood cell (WBC) counting during differential analyses
∙ Digital counter, ideal for use in clinical and research laboratories, comes with a range of features to support precise cell tallying
∙ Eight key digital differential counter is great for simplifying manual reticulocyte counts
∙ Using the WBC maturation chart positioned above the LCD display windows as a guide, press the key corresponding to the cell observed on the microscope slide
∙ Includes an AC adapter and an operation manual
∙ First in class digital operation in a traditional key layout
∙ Eight count keys are easy to push and clearly indicate by sound and touch when a key stroke is entered
∙ Features an audible tone when each 100 cells are counted
∙ There is a one touch reset button  and a subtract button if a mistake is keyed in
Catalog No. NC3302221 Supplier Medicus Health Supplier No. 5267M3
Digital Counting Device is an advanced tool designed for accurate and efficient white blood cell (WBC) counting during differential analyses
∙ Digital counter, ideal for use in clinical and research laboratories, comes with a range of features to support precise cell tallying
∙ Eight key digital differential counter is great for simplifying manual reticulocyte counts
∙ Using the WBC maturation chart positioned above the LCD display windows as a guide, press the key corresponding to the cell observed on the microscope slide
∙ Includes an AC adapter and an operation manual
∙ First in class digital operation in a traditional key layout
∙ Eight count keys are easy to push and clearly indicate by sound and touch when a key stroke is entered
∙ Features an audible tone when each 100 cells are counted
∙ There is a one touch reset button  and a subtract button if a mistake is keyed in