Learn More
Labnet International, Inc Digital Dry Bath, Dual block, 120V
Catalog No. NC1302933
Encompass_Preferred
The Labnet Digital Dry bath incubators are available in single, dual, and four block configurations and provide a broad temperature range up to 150°C, which makes them useful for a variety of applications in molecular biology, histology, clinical, environmental, and industrial laboratories. They have been designed to incorporate the best combination of features including block capacity, dual displays, and an external temperature probe that gives life science researchers the confidence and efficiency they need when working with precious samples.
Catalog No. NC1302933 Supplier Labnet International, Inc Supplier No. D1302
The Labnet Digital Dry bath incubators are available in single, dual, and four block configurations and provide a broad temperature range up to 150°C, which makes them useful for a variety of applications in molecular biology, histology, clinical, environmental, and industrial laboratories. They have been designed to incorporate the best combination of features including block capacity, dual displays, and an external temperature probe that gives life science researchers the confidence and efficiency they need when working with precious samples.