Learn More
Electron Microscopy Sciences Digital Dry Bath
SDP
Catalog No. 501902276
Encompass
Thermally control samples during use with Digital Dry Bath. Utilizes all CoolSafe system chambers. Temperatures can be controlled from 5 degree above ambient to 100 degree C in 1 degree increments to allow for stable operation. Aluminum block evenly transfer heat to tubes for uniform heat transfer. Each unit comes with a block lifter to safely remove hot chambers for easy removal.
Catalog No. 50-190-2276 Supplier Electron Microscopy Sciences Supplier No. 6711910
Thermally control samples during use with Digital Dry Bath. Utilizes all CoolSafe system chambers. Temperatures can be controlled from 5 degree above ambient to 100 degree C in 1 degree increments to allow for stable operation. Aluminum block evenly transfer heat to tubes for uniform heat transfer. Each unit comes with a block lifter to safely remove hot chambers for easy removal.