Learn More
Research Products International Corp Digital Drybath, Double Block Capacity, 150C Max Temperature, 8 x 10 1/2 x 3 1/4 Inches
SDP
Catalog No. 50550234
Encompass
Digital Dry Baths accomodate a variety of interchangeable heating blocks. Ideal for a variety of applications including coagulation studies, denaturing DNA, hybridization, immunoassays, melting agar, PCR, restriction digests, screening antibodies and more. Digital Dry Baths are microprocessor controlled and offer precise, accurate temperature control. Interior block chamber is constructed of stainless steel and provides a uniform transfer of heat, regardless of sample position. Dry Baths are supplied with a block lifter.
Catalog No. 50-550-234 Supplier Research Products International Corp Supplier No. 246201
Digital Dry Baths accomodate a variety of interchangeable heating blocks. Ideal for a variety of applications including coagulation studies, denaturing DNA, hybridization, immunoassays, melting agar, PCR, restriction digests, screening antibodies and more. Digital Dry Baths are microprocessor controlled and offer precise, accurate temperature control. Interior block chamber is constructed of stainless steel and provides a uniform transfer of heat, regardless of sample position. Dry Baths are supplied with a block lifter.