Learn More
Research Products International Corp Digital Dry Bath Incubators, Single Chamber with Single Block, 7 x 10 x 5 Inches
SDP
Catalog No. 501367697
Encompass
State of the art digital dry bath incubators are featured in single and double block styles. Digital temperature control and a removable hinged cover provide the highest level of temperature accuracy and uniformity. Incubators are supplied with either one or two Combination Flip Blocks depending of model. These unique blocks accommodate 14 x 0.5ml and 24 x 2.0ml tubes on one side and six PCR tube strips; 48 x 0.2ml tubes; or one PCR plate on the flip side. Chambers will also accept other standard size blocks with dimensions of 23 x 3 3/4 x 2 inches.
Catalog No. 50-136-7697 Supplier Research Products International Corp Supplier No. 246320
State of the art digital dry bath incubators are featured in single and double block styles. Digital temperature control and a removable hinged cover provide the highest level of temperature accuracy and uniformity. Incubators are supplied with either one or two Combination Flip Blocks depending of model. These unique blocks accommodate 14 x 0.5ml and 24 x 2.0ml tubes on one side and six PCR tube strips; 48 x 0.2ml tubes; or one PCR plate on the flip side. Chambers will also accept other standard size blocks with dimensions of 23 x 3 3/4 x 2 inches.