Learn More
Electron Microscopy Sciences Digital Dry Bath with Cham-3840 and Lifter
SDP
Catalog No. 501902278
Encompass
Thermally control samples during use with Digital Dry Bath. Utilizes all CoolSafe system chambers. Temperatures can be controlled from 5 degree above ambient to 100 degree C in 1 degree increments to allow for stable operation. Aluminum block evenly transfer heat to tubes for uniform heat transfer. Each unit comes with a block lifter to safely remove hot chambers for easy removal.
Catalog No. 50-190-2278 Supplier Electron Microscopy Sciences Supplier No. 6711930
Thermally control samples during use with Digital Dry Bath. Utilizes all CoolSafe system chambers. Temperatures can be controlled from 5 degree above ambient to 100 degree C in 1 degree increments to allow for stable operation. Aluminum block evenly transfer heat to tubes for uniform heat transfer. Each unit comes with a block lifter to safely remove hot chambers for easy removal.