Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Digital Dry Baths/Block Heaters
SCI_ED

Catalog No. S37337 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
1 Block
2 Blocks
Dimensions (L x W x H):
288 x 200 x 100 mm
318 x 200 x 100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Dimensions (L x W x H)
S37337 1 Block 288 x 200 x 100 mm
S37338 2 Blocks 318 x 200 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S37337 Supplier Thermo Scientific™ Supplier No. 88870001/EMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Increase lab versatility with the Thermo Scientific™ digital dry baths/block heaters.

Increase lab versatility with the Thermo Scientific™ digital dry baths, which offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a max temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Required Accessory:
  • Unit must be ordered with block(s)

Warranty and Services

2 Year, Replacement

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID digital
Holds 1 Block
Plug Type US
Relative Humidity <80%
Temperature Range (Metric) Ambient +5° to 130°C
Type Dry Block Heater
Fuse 250V 2.5A
Heating Rate ≤ 20 min. 30° to 130°C
Includes Dry bath, US style plug, block lifter
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 1°C
Timer 0 to 99:59 min or continuous
Warranty 2 Year, Replacement
Dimensions (L x W x H) 288 x 200 x 100 mm
Length (English) 11.3 in.
Length (Metric) 288 mm
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 3.9 in.
Height (Metric) 100 mm
Weight (English) 6.39 lb.
Weight (Metric) 2.9 kg
Voltage 100/120 V
Frequency 50/60 Hz
Show More Show Less

Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.