Learn More
JDM LAB SOLUTIONS LLC Digital Dry Baths DB-03A
Catalog No. NC2385895
Encompass
High-quality heating elements provide excellent temperature uniformityOverheating alarm and automatic shut off featureA wide variety of aluminum tube/plate blocks are availableIndicator light shows when the bath is >60⁰CProgrammable function allows for multiple temperature control points (For models DB-01A, DB-03A, and DB-04A)Set to run continuously or set a timer up to 99 hours 59 minutes (For models DB-01A, DB-03A, and DB-04A) 
Catalog No. NC2385895 Supplier JDM LAB SOLUTIONS LLC Supplier No. DB03A
High-quality heating elements provide excellent temperature uniformityOverheating alarm and automatic shut off featureA wide variety of aluminum tube/plate blocks are availableIndicator light shows when the bath is >60⁰CProgrammable function allows for multiple temperature control points (For models DB-01A, DB-03A, and DB-04A)Set to run continuously or set a timer up to 99 hours 59 minutes (For models DB-01A, DB-03A, and DB-04A) 