Learn More
Hach Company Digital Extension Cable for SC Sensors, 15 m (49 ft) - For Hach SC Digital Controllers
Catalog No. NC3779871 Shop All Hach Company Products
Encompass
The Hach LZX850 digital extension cable provides a 15 m (49 ft) extension for SC digital sensors and controllers, allowing sensors to be positioned away from the controller while maintaining reliable digital communication.
  • Extends digital sensor cable by 15 m (49 ft).
  • Designed for use with SC digital sensors and SC digital controllers.
  • Preserves digital communication between sensor and controller.
  • Inline connection for straightforward installation.
Catalog No. NC3779871 Supplier Hach Company Supplier No. LZX850
The Hach LZX850 digital extension cable provides a 15 m (49 ft) extension for SC digital sensors and controllers, allowing sensors to be positioned away from the controller while maintaining reliable digital communication.
  • Extends digital sensor cable by 15 m (49 ft).
  • Designed for use with SC digital sensors and SC digital controllers.
  • Preserves digital communication between sensor and controller.
  • Inline connection for straightforward installation.