Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Hach Digital Extension Cable, 15 m (50 ft) - for Digital pH/ORP Sensors and SC200 Controller

Catalog No. NC2173620 Shop All Hach Company Products
Encompass

The Hach digital extension cable, 15 m (50 ft), extends the cable length of compatible digital sensors and controllers to allow remote mounting and longer installation runs. The cable is constructed for reliable signal transmission in process environments.

  • Extends sensor cable length by 15 m (50 ft).
  • 4-conductor plus 1 shield cable construction.
  • Rated to 105 °C (221 °F).
  • Recommended maximum single-run cable length: 100 m (328 ft).
  • When used with a termination box, enables transmission distances up to 1,000 m (3,280 ft).

This item is not returnable. View return policy
Catalog No. NC2173620 Supplier Hach Company Supplier No. 5796100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Hach digital extension cable, 15 m (50 ft), extends the cable length of compatible digital sensors and controllers to allow remote mounting and longer installation runs. The cable is constructed for reliable signal transmission in process environments.

  • Extends sensor cable length by 15 m (50 ft).
  • 4-conductor plus 1 shield cable construction.
  • Rated to 105 °C (221 °F).
  • Recommended maximum single-run cable length: 100 m (328 ft).
  • When used with a termination box, enables transmission distances up to 1,000 m (3,280 ft).

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.