Learn More
Hach Company Digital Extension Cable 30m
Catalog No. 5000207445 Shop All Hach Company Products
Encompass
Digital Extension Cable-30 m (100 ft)Digital extension cable is used only with digital sensors or gateways when connecting to any Hach SC digital controller Maximum cable length without Termination Box is 60 m (197 ft ) Maximum cable length with Termination Box (5867000) is 400 m (1312 ft )
Catalog No. 50-002-07445 Supplier Hach Company Supplier No. 5796200
Digital Extension Cable-30 m (100 ft)Digital extension cable is used only with digital sensors or gateways when connecting to any Hach SC digital controller Maximum cable length without Termination Box is 60 m (197 ft ) Maximum cable length with Termination Box (5867000) is 400 m (1312 ft )