Learn More
Hach Company Digital Extension Cable 7.7m
Supplier:  Hach Company 5796000
Encompass
Digital Extension Cable-7 7 m (25 ft)Digital extension cable is used only with digital sensors or gateways when connecting to any Hach SC digital controller Maximum cable length without Termination Box is 60 m (197 ft ) Maximum cable length with Termination Box (5867000) is 400 m (1312 ft )
Catalog No. 50-002-07248