Learn More
AVANTOR MASTERFLEX (VWR LLC) GF Signet 3-8150-1 8150 Field Mount Flow Monitor/Totalizer, Battery Powered
Catalog No. 50631032
Encompass
Features a loop-powered digital flow transmitter, battery-powered digital flow rate monitor and totalizer and line-powered analog/digital flow controller.
Catalog No. 50-631-032 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3248729
Features a loop-powered digital flow transmitter, battery-powered digital flow rate monitor and totalizer and line-powered analog/digital flow controller.