Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Digital Benchtop Gear Pump System, 0.91 mL/rev, 75 psi; 90 to 260 VAC
Catalog No. NC1606817
Encompass
Calibrate, copy, and dispense features produce precise, pulseless batching and dispensing. View pump performance continually. Retrieve displacement and flow values by pump head size. These precision systems are easily operated from the intuitive, multilanguage LCD front panel or optional foot switch 07523-92 for hands-free on/off control. The IP33-rated enclosure protects internal components from damage. Service kits 07144-42 and 74012-52 include: gears, suction shoe, shaft seals, and springs Service kit 74012-22 includes: gears, bushings, and seals
Catalog No. NC1606817 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7401432
Calibrate, copy, and dispense features produce precise, pulseless batching and dispensing. View pump performance continually. Retrieve displacement and flow values by pump head size. These precision systems are easily operated from the intuitive, multilanguage LCD front panel or optional foot switch 07523-92 for hands-free on/off control. The IP33-rated enclosure protects internal components from damage. Service kits 07144-42 and 74012-52 include: gears, suction shoe, shaft seals, and springs Service kit 74012-22 includes: gears, bushings, and seals