Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL HEAT ULTRA S BATH 120V
Catalog No. NC3721474
Encompass_Preferred
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX3800H, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank, Timer: 90 min.
Catalog No. NC3721474 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952318R
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX3800H, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank, Timer: 90 min.