Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL HEAT ULTRASONIC BATH
Catalog No. NC3721482
Encompass_Preferred
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX8800H-E, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Voltage: 230/240V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank
Catalog No. NC3721482 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952838R
Add to Cart
Add to Cart
Request a Quote
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX8800H-E, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Voltage: 230/240V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank