Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL HEATED ULTRASONIC BATH
Catalog No. NC3721473
Encompass_Preferred
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX2800H, Capacity: 2.8L (0.75 gal.), Dimensions Interior: 23 x 14 x 10cm (9.5 x 5.5 x 4 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover, Plastic Housing, Stainless Steel Tank, Timer: 90 min.
Catalog No. NC3721473 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952218R
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX2800H, Capacity: 2.8L (0.75 gal.), Dimensions Interior: 23 x 14 x 10cm (9.5 x 5.5 x 4 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover, Plastic Housing, Stainless Steel Tank, Timer: 90 min.