Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL HEATED ULTRASONIC BATH
Catalog No. NC3721476
Encompass_Preferred
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX5800H, Capacity: 9.5L (2.5 gal.), Dimensions Interior: 29 x 24 x 15cm (11.5 x 9.5 x 6 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank, Timer: 90 min.
Catalog No. NC3721476 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952518R
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX5800H, Capacity: 9.5L (2.5 gal.), Dimensions Interior: 29 x 24 x 15cm (11.5 x 9.5 x 6 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank, Timer: 90 min.