Learn More
Cole-Parmer Cole-Parmer Stuart Digital Hot Plate, 110V
Supplier:  Cole-Parmer 0466123
Encompass_Preferred
This sleek hot plate is simple to use and provides exact heating for all your benchtop needs.It has an intuitive control panel with a unique pad that allows all operations to be set using just your fingertip. An LCD display shows both set and real-time parameters, and is lockable to prevent accidental changes during use. Its optimal heating is produced by an integrated temperature sensor and dual PID control that rapidly heats plate and maintains desired temperature. It has dual temperature control circuits to also ensure temperature stability in case one circuit malfunctions. Its safety features consist of a user-set safety temperature limit through the main control panel, hot warning light indicator, and digital error code functions for easy troubleshooting.
Catalog No. 50-196-0388