Learn More
Research Products International Corp Digital Hot Plate Stirrer, 12x12"" Aluminum Top
SDP
Catalog No. 501976880
Encompass
The EchoTherm Digital Hot Plate Stirrer is designed to be the easiest to use and most accurate fully featured stirring hot plate in the world. Temperature measurement of a solution is accurate to 1% over the entire temperature range using a 100 ohm platinum RTD sensor and measurement system.
Calibration of temperature is electronic, set at the factory and stored in memory. The chassis is cast aluminum designed to keep spills and heat out of the electronics. They are coated with chemically resistant epoxy based paint for durability.
  •      Extremely easy use by anyone in your lab  
  •      Two memory buttons for storing your favorite settings for heater and stirrer for instant, one-button recall and use  
  •      Platinum RTD temperature measurement for precision and accuracy  
Catalog No. 50-197-6880 Supplier Research Products International Corp Supplier No. 150612
The EchoTherm Digital Hot Plate Stirrer is designed to be the easiest to use and most accurate fully featured stirring hot plate in the world. Temperature measurement of a solution is accurate to 1% over the entire temperature range using a 100 ohm platinum RTD sensor and measurement system.
Calibration of temperature is electronic, set at the factory and stored in memory. The chassis is cast aluminum designed to keep spills and heat out of the electronics. They are coated with chemically resistant epoxy based paint for durability.
  •      Extremely easy use by anyone in your lab  
  •      Two memory buttons for storing your favorite settings for heater and stirrer for instant, one-button recall and use  
  •      Platinum RTD temperature measurement for precision and accuracy  