Learn More
Cleatech DIGITAL HYGROMETER
SDP
Catalog No. NC1755420
Encompass_Preferred
Digital Hygrometer / Thermometer (Temp. range: 14° F to 140° F , Humidity range: 15% to 99%RH, Temperature accuracy: ±1.8 F, Humidity accuracy: ±5%, Resolution)
Catalog No. NC1755420 Supplier Cleatech Supplier No. A15MTHTS
Digital Hygrometer / Thermometer (Temp. range: 14° F to 140° F , Humidity range: 15% to 99%RH, Temperature accuracy: ±1.8 F, Humidity accuracy: ±5%, Resolution)