Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Digital Infant Scale
SDP

Catalog No. 22445289 Shop All Mopec Products
Change view
Click to view available options
Readability:
0.01 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Readability
22-445-289 0.01 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-289 Supplier Mopec™ Supplier No. BB064
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This infant scale provides digital readout in pounds/ounces or kilograms/grams. Scale is battery operated and features color-coded keypad with instructions and lock-in weight display.

  • Removable white molded plastic tray has raised sides for enhanced safety
  • Easy to wipe clean
  • Includes standard serial output for use with computer or printer

Specifications

Capacity 44 lb.
Display Type LCD
Readability 0.01 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.