Learn More
Braintree Scientific Digital Lab Std Stereotaxic Instrument w/45 degree ear bars
SDP
Catalog No. 501953996
Encompass
Digital Lab Standard™ features a sealed electronic sensor, highly immune to interference and environmental conditions. Movements are monitored by a large, easy-to-read LED display module and are accurate to 10uL. A zeroing function allows the user to target specific coordinates in the brain for injection, implantation, etc. Calculation of distance measurements via the vernier scales is thus no longer necessary...saving the user valuable time and improving accuracy. **Sold for research purposes only and not for human use**
Catalog No. 50-195-3996 Supplier Braintree Scientific Supplier No. 51950
Add to Cart
Add to Cart
Request a Quote
Digital Lab Standard™ features a sealed electronic sensor, highly immune to interference and environmental conditions. Movements are monitored by a large, easy-to-read LED display module and are accurate to 10uL. A zeroing function allows the user to target specific coordinates in the brain for injection, implantation, etc. Calculation of distance measurements via the vernier scales is thus no longer necessary...saving the user valuable time and improving accuracy. **Sold for research purposes only and not for human use**