Learn More
Research Products International Corp Digital Laboratory Incubator, Acrylic See Through Door, 220 volt, 0.7 Cubic Feet Capacity
SDP
Catalog No. 501977011
Encompass
Digital accuracy and performance incubators offered at an affordable price. Full PID microprocessor controller maintains temperature accuracy within 1°C. Innovative built in tracking system stores maximum temperature deviation from set point in 0.1° increments. This feature acts as a paperless chart recorder to confirm stability or indicate any system malfunction. Supplied complete with one fixed and one adjustable chrome plated shelf.
Additional features include:
  • Digital read out in 0.1°C or F
  • Self diagnostic software
  • div>WARNING  This product can expose you to chemicals including Diethylhexyl Phthalate (DEHP), which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7011 Supplier Research Products International Corp Supplier No. 210014220
Digital accuracy and performance incubators offered at an affordable price. Full PID microprocessor controller maintains temperature accuracy within 1°C. Innovative built in tracking system stores maximum temperature deviation from set point in 0.1° increments. This feature acts as a paperless chart recorder to confirm stability or indicate any system malfunction. Supplied complete with one fixed and one adjustable chrome plated shelf.
Additional features include:
  • Digital read out in 0.1°C or F
  • Self diagnostic software
  • div>WARNING  This product can expose you to chemicals including Diethylhexyl Phthalate (DEHP), which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.