Learn More
World Precision Instrument Digital Manipulator Arm, 3-Axes, Add-On, Left handed
SDP
Catalog No. 502539101
Encompass
You should consider an upgrade if you currently own a vernier model stereotaxic. We can upgrade most major stereotaxic brands. Upgrading your existing stereotaxic to a digital model will reduce the risk of human error and increase accuracy (10-rather than 100-micron resolution). The large LED display is also useful for surgeries performed in low light, or constrained environments, where reading the vernier scale is difficult.
Catalog No. 50-253-9101 Supplier World Precision Instrument Supplier No. 505356
You should consider an upgrade if you currently own a vernier model stereotaxic. We can upgrade most major stereotaxic brands. Upgrading your existing stereotaxic to a digital model will reduce the risk of human error and increase accuracy (10-rather than 100-micron resolution). The large LED display is also useful for surgeries performed in low light, or constrained environments, where reading the vernier scale is difficult.