Learn More
World Precision Instrument Digital Manipulator Arm, 3-Axes, Conversion, Right handed
SDP
Catalog No. 502539095
Encompass
You should consider an upgrade if you currently own a vernier model WPI Premium stereotaxic frame.  Upgrading your existing WPI Premium stereotaxic frame to a digital model will reduce the risk of human error and increase accuracy (10-micron from100-micron resolution). The large LED display is also useful for surgeries performed in low light, or constrained environments where reading the vernier scale is difficult.
Catalog No. 50-253-9095 Supplier World Precision Instrument Supplier No. 505362
You should consider an upgrade if you currently own a vernier model WPI Premium stereotaxic frame.  Upgrading your existing WPI Premium stereotaxic frame to a digital model will reduce the risk of human error and increase accuracy (10-micron from100-micron resolution). The large LED display is also useful for surgeries performed in low light, or constrained environments where reading the vernier scale is difficult.