Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digital Meters
SCI_ED

Catalog No. S02336 Shop All Luster Leaf Products, Inc. Products
Change view
Click to view available options
Type:
Digital Moisture Meter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S02336 Digital Moisture Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02336 Supplier Luster Leaf Products, Inc. Supplier No. 1825

Designed for simple, accurate test readings.

Digital Moisture Meter

  • New digital results from 0 to 9.9
  • Printed instructions and extensive plant list packed with the meter

Digital pH Meter
  • New digital results from 3.5 to 9.0
  • Printed instructions and plant pH preference list packed with the meter

Digital PLUS Moisture Meter
  • New digital results from 0 to 9.9
  • Extensive plant list with preferred soil moisture levels and special plant needs built in to meter database

Digital PLUS Soil pH Meter
  • New digital results from 3.5 to 9.0
  • Plant pH preference list is built in to the meter database

Note: Probe should not be left in contact with moisture for extended periods of time.

Specifications

Includes Printed instructions and extensive plant list packed with the meter
For Use With (Application) Moisture
Detection Range 0 to 9.9 pH
Type Digital Moisture Meter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.