Learn More
Uline Mitutoyo Digital Micrometer 0-1""
Catalog No. NC2239452
Encompass
Mitutoyo Digital MicrometerSimple one-hand operation for precise, reliable measuring.For rigid materials like sheet metal, boards, tubing and small parts.Easy-to-read digital display. Inch/millimeter convertible.Enamel finished cast iron frame.Friction thimble adjustment.Zero function; auto on/off saves battery.
This item is not returnable. View return policy
Catalog No. NC2239452 Supplier Uline Supplier No. H2780
Mitutoyo Digital MicrometerSimple one-hand operation for precise, reliable measuring.For rigid materials like sheet metal, boards, tubing and small parts.Easy-to-read digital display. Inch/millimeter convertible.Enamel finished cast iron frame.Friction thimble adjustment.Zero function; auto on/off saves battery.