Learn More
Grainger Mitutoyo MDC Lite Digital Outside Micrometer, 0 in. to 1 in., 1 ea.
Catalog No. NC2952064
Encompass_Preferred
This digital outside micrometer is designed for precise length measurements up to 25 mm (1 in.). It features an easy-to-read LCD display for both metric and imperial units and is ideal for applications where rapid, consistent measurement is required without the need for external data transmission.
  • Digital LCD display with inch/mm conversion
  • Carbide-tipped measuring faces for durability
  • Constant measuring force via ratchet stop
  • IP65 protection against dust and water
  • Includes a plastic fitted case and certificate of inspection
  • Automatic power-off function after 20 minutes of idle time
  • Powered by an SR44 battery with approximately 2.4-year battery life
Catalog No. NC2952064 Supplier Grainger Supplier No. 1ARD6
This digital outside micrometer is designed for precise length measurements up to 25 mm (1 in.). It features an easy-to-read LCD display for both metric and imperial units and is ideal for applications where rapid, consistent measurement is required without the need for external data transmission.
  • Digital LCD display with inch/mm conversion
  • Carbide-tipped measuring faces for durability
  • Constant measuring force via ratchet stop
  • IP65 protection against dust and water
  • Includes a plastic fitted case and certificate of inspection
  • Automatic power-off function after 20 minutes of idle time
  • Powered by an SR44 battery with approximately 2.4-year battery life