Learn More
Surgical Design™ Digital Micrometer
Catalog No. 21124918 Shop All Surgical Design Products
Catalog No. 21-124-918 Supplier Surgical Design™ Supplier No. MT002
Add to Cart
Add to Cart
Digital micrometer delivers ultra-accurate thickness and diameter measurements for laboratory, engineering, industrial, and quality-control applications.
  • High-resolution digital display for fast, accurate, and easy-to-read measurements
  • Precision spindle and anvil deliver consistent, repeatable micrometer readings
  • Durable construction engineered for reliability in laboratory and industrial use
  • Smooth ratchet mechanism prevents over-tightening and ensures measurement accuracy
  • Instant unit conversion (mm/in.) for workflow flexibility
  • Ideal for lab work, machining, quality control, and engineering applications