Learn More
Braintree Scientific Digital Microscope/Camera Kit,DinoCapture software & bracket
SDP
Catalog No. 501955486
Encompass
Portable Digital Microscope/CameraEquip your surgery platform with a portable digital microscope/camera Ideal for small rodent surgery, demonstrations, dissections and much more! Features a 2.0 USB connection eliminating the need for interface cards or expensive software. Easy and convenient to load and use DinoCapture application software and users manual are included for collection and processing of photos. Resolution: 640 x 480 pixels Magnification: 10~200x continuous Sensor: 1/4" Color CMOS Video frame rate: up to 30 frame-per second Illumination: built-in 8 white LEDs CD Content: DinoCapture application software, users manual Automatic or manual color balance and brightness control System Requirements: Compatible Windows 2000/XP, Vista and most MAC OS Intel Pentium III 1 GHz processor or faster 256 MB RAM CD-ROM drive 16-bit color display monitor or higher Available hi-speed USB 2.0 port **Sold for research purposes only and not for human use**
Catalog No. 50-195-5486 Supplier Braintree Scientific Supplier No. PDMC
Portable Digital Microscope/CameraEquip your surgery platform with a portable digital microscope/camera Ideal for small rodent surgery, demonstrations, dissections and much more! Features a 2.0 USB connection eliminating the need for interface cards or expensive software. Easy and convenient to load and use DinoCapture application software and users manual are included for collection and processing of photos. Resolution: 640 x 480 pixels Magnification: 10~200x continuous Sensor: 1/4" Color CMOS Video frame rate: up to 30 frame-per second Illumination: built-in 8 white LEDs CD Content: DinoCapture application software, users manual Automatic or manual color balance and brightness control System Requirements: Compatible Windows 2000/XP, Vista and most MAC OS Intel Pentium III 1 GHz processor or faster 256 MB RAM CD-ROM drive 16-bit color display monitor or higher Available hi-speed USB 2.0 port **Sold for research purposes only and not for human use**