Learn More
National Optical™ Digital Microscope with Camera
SCI_ED
Catalog No. S012583
Change view
Click to view available options
Objective Types:
DIN Achromatic
Catalog No. Objective Types
S012583 DIN Achromatic
1 options
Catalog No. S012583 Supplier National Optical™ Supplier No. DC4212
Add to Cart
Add to Cart
Now with built-in digital 5.0 MP camera, this digital microscope offers a new standard of precision optics, highly engineered mechanics, and very rugged construction. Reverse position objective turret permits easier changing and positioning of specimen slides. Preferred by teachers, the corded LED illumination system provides optimum image brightness, virtually no bulb heat, and has a 50,000 hour rated bulb life.
Now with built-in digital 5.0 MP camera, the DC4-212 offers a new standard of precision optics, highly engineered mechanics and very rugged construction. Objective turret (nosepiece) is in reverse position to permit easier changing and positioning of specimen slides. Preferred by teachers, the corded LED illumination system provides optimum image brightness, virtually no bulb heat and has a 50,000-hour rated bulb life.
OPTICAL SYSTEM
  • Widefield 10x eyepiece with pointer. Accepts optional #965-160 eyepiece reticle.
  • 45° inclined monocular viewing head rotates 360° for easy sharing by more than one student.
  • DIN standard 4x (0.10 N.A.), 10x (0.25 N.A.), 40xR (0.65 N.A.) and 100xR (1.25 N.A.) objectives are supplied for 40x, 100x, 400x and 1000x magnification. All are achromatic, parfocalled, parcentered and color-coded.
  • 4-hole objective turret, reverse position, ball-bearing mounted for smooth, precise changes in magnification.
ILLUMINATION
  • Built-in corded LED illumination, delivers greater light and longer life than standard 20-watt tungsten bulb.
  • Virtually no base heat and a 50,000-hour rated bulb.
  • Base mounted on/off switch, heavy-duty 3-wire grounded electrical system and plug.
  • 0.5A time delay fuse.
  • Rheostat intensity control.
FOCUSING
  • Separate low position coarse and fine focusing controls.
  • Stage moves up or down, eyepiece and body remain fixed.
  • Slip clutch prevents damage to focusing system.
  • Tension adjustment eliminates stage drift.
  • Safety rack stop prevents damage to slides and objective lenses
STAGE
  • Large 120 x 120 mm stage, with professional quality, low profile. mechanical specimen holder with low-position coaxial x-y controls.
  • 1.25 N.A. Abbe condenser with spiral focusing mount, iris diaphragm, swing-out filter holder, blue filter supplied.
CAMERA SPECIFICATIONS
  • 5.0MP
  • Sensor Type: CMOS
  • Calibration slide and software included.
FRAME
  • Rugged cast metal, gray enamel finish.
Specifications
Eyepiece 10X/18 mm, with pointer
Head Style Monocular
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Objective Types DIN Achromatic
Product Type Digital Imaging Microscope
Voltage 110/220 V
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov