Learn More
Andwin Scientific DIGITAL MINI INCUBATOR 1/EA
SDP
Catalog No. 501126011
Encompass_Preferred
The 115V digital mini incubator is ideal for microbiology and hematology applications. It offers incremental temperature adjustments, transparent Plexiglas door for visibility, epoxy-coated steel walls and a capacity of 10 liters.
Catalog No. 50-112-6011 Supplier Andwin Scientific Supplier No. 10055006
The 115V digital mini incubator is ideal for microbiology and hematology applications. It offers incremental temperature adjustments, transparent Plexiglas door for visibility, epoxy-coated steel walls and a capacity of 10 liters.