Learn More
JDM LAB SOLUTIONS LLC Digital Mini Water Bath WB-01A
Catalog No. NC2385925
Encompass
Microprocessor PID control with digital displayExcellent temperature uniformityOverheating alarm and automatic shut offDry run protection with audible alarm to avoid damage if the water level is too lowIndicator light illuminates when temperature >60°CVarious tube holders are available to order separately
Models WB-01A is available with a programmable feature.
Catalog No. NC2385925 Supplier JDM LAB SOLUTIONS LLC Supplier No. WB01A
Microprocessor PID control with digital displayExcellent temperature uniformityOverheating alarm and automatic shut offDry run protection with audible alarm to avoid damage if the water level is too lowIndicator light illuminates when temperature >60°CVarious tube holders are available to order separately
Models WB-01A is available with a programmable feature.