Learn More
National Optical Digital Monocular Microscope with 5.0MP Built-in Camera
SCI_ED
Catalog No. S81783
Change view
Click to view available options
Magnification Power:
4X, 10X, 40X, 100X
Catalog No. Magnification Power
S81783 4X, 10X, 40X, 100X
1 options
Catalog No. S81783 Supplier National Optical Supplier No. DC128
Add to Cart
Add to Cart
This compound microscope and digital camera combination includes a 100X oil immersion lens, 1.25 N.A. Abbe condenser, and a rechargeable LED illumination system.
This compound microscope and digital camera combination includes a 100x oil immersion lens, 1.25 N.A. Abbe condenser, and a rechargeable LED illumination system.  Motic Images software included. Use with Windows 7 and above.  Mac OSX Compatible. 
OPTICAL SYSTEM
  • Widefield 10x eyepiece with pointer.
  • 45° inclined monocular viewing head.
  • DIN standard 4x (0.10 N.A.), 10x (0.25 N.A.) and 40xR (0.65 N.A.) objectives are supplied for 40x, 100x and 400x magnification. Also included is a 100xR (1.25 N.A.) oil immersion lens for 1000x magnification.  All lenses are achromatic, parfocalled, parcentered and color-coded.
  • 4-hole objective turret with click-stops, ball-bearing mounted for smooth, precise changes in magnification.
ILLUMINATION
  • LED illumination rated up to 50,000 hours life, with light output equal to a 20 watt tungsten bulb, totally heat free.
  • Rechargeable AA nickel metal hydride batteries can be recharged up to 500 times, with each 8 hour charge providing up to 50 hours of operation.
  • Included recharger shuts off when batteries are fully charged, eliminating the danger of overcharging and shortening battery life.  Microscope can be used during recharging, or recharger can remain plugged-in for conventional use.
  • Rheostat light intensity control with separate on/off switch.
FOCUSING
  • Separate low position coarse and fine focusing controls.
  • Stage moves up or down, eyepiece and body remain fixed.
  • Slip clutch prevents damage to focusing system.
  • Tension adjustment eliminates stage drift.
  • Safety rack stop prevents damage to slides and objective lenses.
STAGE
  • Large 120 mm x 120 mm (4-3/4 in. x 4-3/4 in.) graduated mechanical stage, with a professional quality low profile specimen holder and low-position coaxial x-y controls.
  • 1.25 N.A. Abbe condenser with spiral focusing mount, iris diaphragm, built-in swing-out frosted glass filter.
Specifications
Magnification Power 4X, 10X, 40X, 100X
Objective Types DIN, achromatic, 4x, 10x, 40xR and 100xR
Nosepiece 4-hole, click stop, ball-bearing mounted
Electrical Requirements 3 rechargeable AA nickel metal hydrate
Condenser Type 1.25 N.A. Abbe condenser with spiral focusing mount and iris diaphragm
Model DC-128
Head Style 45-deg. inclined monocular viewing head
Camera Type 5.0MP digital
Lighting Type LED
Eyepiece 10x, 18mm, with pointer
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov