Learn More
World Precision Instrument WPI Ultra Precise Digital Dual Mouse Stereotaxic Instrument
SDP
Catalog No. 502539122
Encompass
WPI's Dual Digital Ultra Precise Mouse Stereotaxic Instrument is specifically utilized for supporting surgical-based mouse-model focused laboratory research. This model frame is ideal for researchers in need of a versatile, reliable instrument specific to fine-tuned movements, necessary in highly placement-sensitive procedures. The dual capability allows for ease of bilateral placement of probes, tet/electrodes/implant arrays, cannula, as well as low-volume fluid delivery. The ability to have both digital visibility of stereotaxic coordinates paired with enhanced precision permits greater ability of validation, critical for use in publications and repeatability of protocols.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9122 Supplier World Precision Instrument Supplier No. 505322
Add to Cart
Add to Cart
Request a Quote
WPI's Dual Digital Ultra Precise Mouse Stereotaxic Instrument is specifically utilized for supporting surgical-based mouse-model focused laboratory research. This model frame is ideal for researchers in need of a versatile, reliable instrument specific to fine-tuned movements, necessary in highly placement-sensitive procedures. The dual capability allows for ease of bilateral placement of probes, tet/electrodes/implant arrays, cannula, as well as low-volume fluid delivery. The ability to have both digital visibility of stereotaxic coordinates paired with enhanced precision permits greater ability of validation, critical for use in publications and repeatability of protocols.
View our Stereotaxic Frame Compatibility Chart