Learn More
Bellco Glass, Inc. The Bellco Digital Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixin
SDP
Catalog No. 501946209
Encompass_Preferred
The Bellco Digital Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixin
Catalog No. 50-194-6209 Supplier Bellco Glass, Inc. Supplier No. 777410115
The Bellco Digital Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixin