Learn More
Bellco Glass, Inc. The Bellco Digital Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixin
SDP
Supplier:  Bellco Glass, Inc. 777410230
Encompass_Preferred
The Bellco Digital Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixin
Catalog No. 50-194-6210