Learn More
Andwin Scientific Digital Constant Temperature Oil Bath 5L 200-230V
SDP
Catalog No. NC1769329
Encompass_Preferred
Large capacity 5L constant temperature oil bath with internal diameter of φ240 mm. Designed with flat-shaped bath with no heater or sensor inside for ease in cleaning. Temperature range: ambient +10~180°C. 200-230V.
Catalog No. NC1769329 Supplier Andwin Scientific Supplier No. BO312A
Large capacity 5L constant temperature oil bath with internal diameter of φ240 mm. Designed with flat-shaped bath with no heater or sensor inside for ease in cleaning. Temperature range: ambient +10~180°C. 200-230V.