Learn More
Yamato Scientific America, Inc. Yamato 7L Constant Temperature Oil Bath, 115V/220V.
Catalog No. 501898177 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Easy to use, compact 7L oil bath with digital setting and display. Protected oil tank prevents burns caused by contact. Equipped with a drain. Temperature range: ambient +5~180°C.
Catalog No. 50-189-8177 Supplier Yamato Scientific America, Inc. Supplier No. BO601115V
Easy to use, compact 7L oil bath with digital setting and display. Protected oil tank prevents burns caused by contact. Equipped with a drain. Temperature range: ambient +5~180°C.