Learn More
Research Products International Corp Orbit P4 Digital Shaker with Platform for 4 Microplates
SDP
Catalog No. 50998281
Encompass
The small Orbit Digital Shaker features variable speed across a broad range, making it useful for a variety of applications. Both time of operation and shaker speed are set and displayed digitally, with their values shown on the large 3 digit LED. Parameters are selected with an easy turn knob. A sturdy base encloses the motor for quiet, vibration free operation. Model can be used in temperature controlled environments. Their compact size makes them ideal for incubator use.
Catalog No. 50-998-281 Supplier Research Products International Corp Supplier No. 192045
The small Orbit Digital Shaker features variable speed across a broad range, making it useful for a variety of applications. Both time of operation and shaker speed are set and displayed digitally, with their values shown on the large 3 digit LED. Parameters are selected with an easy turn knob. A sturdy base encloses the motor for quiet, vibration free operation. Model can be used in temperature controlled environments. Their compact size makes them ideal for incubator use.