Learn More
Biotang Inc Digital Orbit Shaker is a powerful variable speed shaker which provides efficient linear motion. max shaking weight of 7.5kg, 9 type of platforms selective for flasks, .PC control via RS232 interface, available to control all measurement
Catalog No. 507514955 Shop All BIOTANG Inc Products
Encompass_Preferred
Digital Orbit Shaker is a powerful variable speed shaker which provides efficient linear motion. max shaking weight of 7.5kg, 9 type of platforms selective for flasks, .PC control via RS232 interface, available to control all measurement
Catalog No. 50-751-4955 Supplier Biotang Inc Supplier No. H70221
Digital Orbit Shaker is a powerful variable speed shaker which provides efficient linear motion. max shaking weight of 7.5kg, 9 type of platforms selective for flasks, .PC control via RS232 interface, available to control all measurement